

4-Chlorophenyl-[3-(trifluoromethyl)phenoxy]acetic acid
Catalog # APOSPC450575-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:330.686 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
:GHS09
CAS Number:4687-08-5
Formula:C₁₅H₁₀ClF₃O₃
MDL Number:MFCD09752899