(1-Ethoxy-2,2,2-trifluoroethoxy) triethylsilane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1254592-76-1
:
Formula:C₁₀H₂₁F₃O₂Si
MDL Number:MFCD30730296
:258.353 g/mol
Boiling Pt:66…67 °C (8 mm Hg)
Storage Temperature:Ambient
Frequently Bought Together