3-(2,2,3,3-Tetrafluoropropoxy)propyl-1-ene carbonate
Catalog # APOSPC450494-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
:
:232.13 g/mol
Storage Temperature:Ambient
CAS Number:879496-46-5
Formula:C₇H₈F₄O₄
MDL Number:MFCD28901315