
4-Chlorophenyl trifluoromethanesulphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29540-84-9
:
Formula:C₇H₄ClF₃O₃S
MDL Number:MFCD07784323
Flash Pt:>110 °C
:260.618 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together