Order Entry
Italy
ContactUsLinkComponent
1H,1H,7H-Perfluoroheptyl 2,2,3,4,4,4-hexafluorobutyl carbonate
1H,1H,7H-Perfluoroheptyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Catalog # APOSPC450362-25G
Supplier:  Apollo Scientific
CAS Number:  
1H,1H,7H-Perfluoroheptyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Catalog # APOSPC450362-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :540.145 g/mol
Storage Temperature:Ambient
CAS Number:1980045-16-6
Formula:C₁₂H₆F₁₈O₃
MDL Number:MFCD28334355