Order Entry
Italy
ContactUsLinkComponent
Allyl 2,2-difluoroethyl ether
Allyl 2,2-difluoroethyl ether
Catalog # APOSPC450065-1G
Supplier:  Apollo Scientific
CAS Number:  
Allyl 2,2-difluoroethyl ether
Catalog # APOSPC450065-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :122.113 g/mol
Storage Temperature:Ambient
CAS Number:111512-41-5
Formula:C₅H₈F₂O
MDL Number:MFCD19442068