2,2-Difluoro-2-(2-methylphenyl)ethan-1-ol
Catalog # APOSPC450038-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:172.172 g/mol
Storage Temperature:Ambient
CAS Number:1365808-64-5
Formula:C₉H₁₀F₂O
MDL Number:MFCD22124009