tert-Butyl 4-(trifluoromethyl)-5,6-dihydropyridine-1(2H)-carboxylate 95%
Catalog # APOSPC448212-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:251.245 g/mol
Storage Temperature:Ambient
CAS Number:291289-18-4
Formula:C₁₁H₁₆F₃NO₂
MDL Number:MFCD28967983