tert-Butyl 5-bromo-4-fluoro-5,6-dihydropyridine-1(2H)-carboxylate 95%
Catalog # APOSPC448211-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:280.134 g/mol
Storage Temperature:Ambient
CAS Number:1881331-83-4
Formula:C₁₀H₁₅BrFNO₂
MDL Number:MFCD28975649