Order Entry
Italy
ContactUsLinkComponent
2-[(Dimethylamino)methylene]-5-(4-fluorophenyl)-1,3-cyclohexanedione 95%
2-[(Dimethylamino)methylene]-5-(4-fluorophenyl)-1,3-cyclohexanedione 95%
Catalog # APOSPC448208-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-[(Dimethylamino)methylene]-5-(4-fluorophenyl)-1,3-cyclohexanedione 95%
Catalog # APOSPC448208-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :261.291 g/mol
Storage Temperature:Ambient
CAS Number:1030419-83-0
Formula:C₁₅H₁₆FNO₂
MDL Number:MFCD13811664