Ethyl 4-fluoropiperidine-4-carboxylate hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:845909-49-1
:
Formula:C₈H₁₄FNO₂·ClH
MDL Number:MFCD09880187
:211.662 g/mol
Storage Temperature:Ambient
Frequently Bought Together