Order Entry
Italy
ContactUsLinkComponent
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethylamine hydrochloride 95+%
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethylamine hydrochloride 95+%
Catalog # APOSPC446176-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]ethylamine hydrochloride 95+%
Catalog # APOSPC446176-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :261.072 g/mol
Storage Temperature:Ambient
Formula:C₈H₈ClF₃N₂·ClH
MDL Number:MFCD31714214