Order Entry
Italy
ContactUsLinkComponent
N-Benzyl-2-(6-morpholin-4-yl-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetamide
N-Benzyl-2-(6-morpholin-4-yl-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetamide
Catalog # APOSPC446080-100MG
Supplier:  Apollo Scientific
CAS Number:  
N-Benzyl-2-(6-morpholin-4-yl-4-trifluoromethyl-pyridin-2-ylsulphanyl)-acetamide
Catalog # APOSPC446080-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :411.441 g/mol
Storage Temperature:Ambient
CAS Number:1431555-23-5
Formula:C₁₉H₂₀F₃N₃O₂S
MDL Number:MFCD24386179