(4-Chloro-phenyl)-[4-(3-chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazin-1-yl]-methanone
Catalog # APOSPC446040-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:418.24 g/mol
Storage Temperature:Ambient
CAS Number:1311278-56-4
Formula:C₁₈H₁₆Cl₂F₃N₃O
MDL Number:MFCD19981201