
Perfluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate 97+%
Catalog # APOSPC4415-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:441.272 g/mol
Melting Pt:91…93 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:898289-52-6
Formula:C₁₈H₁₁F₈NO₃
MDL Number:MFCD09025890