Order Entry
Italy
ContactUsLinkComponent
2-(Trifluoromethylthio)phenylacetic acid 98%
2-(Trifluoromethylthio)phenylacetic acid 98%
Catalog # APOSPC4324-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(Trifluoromethylthio)phenylacetic acid 98%
Catalog # APOSPC4324-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :236.211 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:239080-01-4
Formula:C₉H₇F₃O₂S
MDL Number:MFCD00236349