Ethyl 4-chloro-7-fluoro-6-methoxyquinoline-3-carboxylate
Catalog # APOSPC430569-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Tipo di conf.:Bottle
- Conf:250 mg
- Seq:2590
Specifications
About this item
:
:283.683 g/mol
Storage Temperature:Ambiente
CAS Number:311346-71-1
Formula:C₁₃H₁₁ClFNO₃
MDL Number:MFCD16036879