Order Entry
Italy
ContactUsLinkComponent
1-(2-Bromo-4-fluorobenzyl)-6,7-dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one
1-(2-Bromo-4-fluorobenzyl)-6,7-dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one
Catalog # APOSPC430565-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Bromo-4-fluorobenzyl)-6,7-dihydro-1H-pyrrolo[3,2-c]pyridin-4(5H)-one
Catalog # APOSPC430565-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :323.16 g/mol
Storage Temperature:Ambient
CAS Number:1206969-77-8
Formula:C₁₄H₁₂BrFN₂O
MDL Number:MFCD14585354