7-Fluoro-1,4-dihydro-6-methoxy-4-oxoquinoline-3-carboxylic acid
Catalog # APOSPC430540-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:237.184 g/mol
Storage Temperature:Ambient
CAS Number:622369-36-2
Formula:C₁₁H₈FNO₄
MDL Number:MFCD18157741