Order Entry
Italy
ContactUsLinkComponent
tert-Butyl 9-oxo-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
tert-Butyl 9-oxo-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
Catalog # APOSPC430344-1G
Supplier:  Apollo Scientific
CAS Number:  
tert-Butyl 9-oxo-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
Catalog # APOSPC430344-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :336.307 g/mol
Storage Temperature:Ambient
CAS Number:1823420-46-7
Formula:C₁₄H₁₉F₃N₂O₄
MDL Number:MFCD28016261