9-(4-Bromobutyl)-N-(2,2,2-trifluoroethyl)-9H-fluorene-9-carboxamide
Catalog # APOSPC430251-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:426.27 g/mol
Storage Temperature:Ambient
CAS Number:182438-98-8
Formula:C₂₀H₁₉BrF₃NO
MDL Number:MFCD27978674