Order Entry
Italy
ContactUsLinkComponent
Diethyl 2,2-difluoropentanedioate
Diethyl 2,2-difluoropentanedioate
Catalog # APOSPC430245-1G
Supplier:  Apollo Scientific
CAS Number:  
Diethyl 2,2-difluoropentanedioate
Catalog # APOSPC430245-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :224.202 g/mol
Storage Temperature:Ambient
CAS Number:428-97-7
Formula:C₉H₁₄F₂O₄
MDL Number:MFCD24386313