Order Entry
Italy
ContactUsLinkComponent
2,3-Difluoro-4-nitrophenol
2,3-Difluoro-4-nitrophenol
Catalog # APOSPC430167-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Difluoro-4-nitrophenol
Catalog # APOSPC430167-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.09 g/mol
Storage Temperature:Ambient
CAS Number:123173-60-4
Formula:C₆H₃F₂NO₃
MDL Number:MFCD13185572