Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Caesium perfluoroheptanoate
Caesium perfluoroheptanoate 5 mM in water/acetonitrile (1:1)
Caesium perfluoroheptanoate 5 mM in water/acetonitrile (1:1)
Caesium perfluoroheptanoate 5 mM in water/acetonitrile (1:1)
APOSPC4289-10ML
:
Apollo Scientific
:
Caesium perfluoroheptanoate 5 mM in water/acetonitrile (1:1)
APOSPC4289-10ML
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
10 mL
Seq:
2275
:
:
495.958 g/mol
:
Ambient
:
171198-24-6
:
C₇F₁₃O₂·Cs
:
MFCD04038276