Order Entry
Italy
ContactUsLinkComponent
2-Isocyanato-1-methoxy-4-(trifluoromethyl)benzene 98%
2-Isocyanato-1-methoxy-4-(trifluoromethyl)benzene 98%
Catalog # APOSPC421207-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Isocyanato-1-methoxy-4-(trifluoromethyl)benzene 98%
Catalog # APOSPC421207-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :217.145 g/mol
Storage Temperature:Ambient
CAS Number:16588-75-3
Formula:C₉H₆F₃NO₂
MDL Number:MFCD07644613