Order Entry
Italy
ContactUsLinkComponent
2-(Trifluoromethyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine 95+%
Irritant
2-(Trifluoromethyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine 95+%
Catalog # APOSPC421202-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Trifluoromethyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine 95+%
Catalog # APOSPC421202-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :189.138 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:905274-04-6
Formula:C₇H₆F₃N₃
MDL Number:MFCD09954780