Order Entry
Italy
ContactUsLinkComponent
4'-Fluoro-2-(4-fluorophenyl)acetophenone
Irritant
4'-Fluoro-2-(4-fluorophenyl)acetophenone
Catalog # APOSPC421013-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-Fluoro-2-(4-fluorophenyl)acetophenone
Catalog # APOSPC421013-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.225 g/mol
Boiling Pt:178…180 °C (15 mm Hg)
Melting Pt:94…98 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:366-68-7
Formula:C₁₄H₁₀F₂O
MDL Number:MFCD00055468