Order Entry
Italy
ContactUsLinkComponent
2-(3,4-Difluorophenyl)-4'-methylacetophenone
2-(3,4-Difluorophenyl)-4'-methylacetophenone
Catalog # APOSPC421005-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3,4-Difluorophenyl)-4'-methylacetophenone
Catalog # APOSPC421005-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :246.252 g/mol
Storage Temperature:Ambient
CAS Number:1504609-61-3
Formula:C₁₅H₁₂F₂O
MDL Number:MFCD23417260