
2-Methyl-4-(2,2,2-trifluoroethoxy)aniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1431962-93-4
:
Formula:C₉H₁₀F₃NO·ClH
MDL Number:MFCD25371225
:241.638 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together