
1-(Difluoromethyl)-4-iodo-3-methyl-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1217862-89-9
:
Formula:C₅H₅F₂IN₂
MDL Number:MFCD15146436
:258.008 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together