Order Entry
Italy
ContactUsLinkComponent
2-(3,3-Difluoroazetidin-1-yl)ethan-1-amine
2-(3,3-Difluoroazetidin-1-yl)ethan-1-amine
Catalog # APOSPC411267-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3,3-Difluoroazetidin-1-yl)ethan-1-amine
Catalog # APOSPC411267-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :136.143 g/mol
Storage Temperature:Ambient
CAS Number:2045189-46-4
Formula:C₅H₁₀F₂N₂
MDL Number:MFCD29994892