Order Entry
Italy
ContactUsLinkComponent
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-4,4,4-trifluorobutane-1,3-dione
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-4,4,4-trifluorobutane-1,3-dione
Catalog # APOSPC410376-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(1-Ethyl-3-methyl-1H-pyrazol-4-yl)-4,4,4-trifluorobutane-1,3-dione
Catalog # APOSPC410376-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :248.202 g/mol
Storage Temperature:Ambient
CAS Number:1006322-30-0
Formula:C₁₀H₁₁F₃N₂O₂
MDL Number:MFCD03419774