Order Entry
Italy
ContactUsLinkComponent
 
3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
Catalog # APOSPC410374-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[1'-Ethyl-5-(trifluoromethyl)-1H,1'H-3,4'-bipyrazol-1-yl]propanenitrile
Catalog # APOSPC410374-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.252 g/mol
Storage Temperature:Ambient
CAS Number:1006320-21-3
Formula:C₁₂H₁₂F₃N₅
MDL Number:MFCD08558461