Order Entry
Italy
ContactUsLinkComponent
1-[(2,4-Difluorophenoxy)methyl]-3,5-dimethyl-4-nitro-1H-pyrazole
1-[(2,4-Difluorophenoxy)methyl]-3,5-dimethyl-4-nitro-1H-pyrazole
Catalog # APOSPC410287-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(2,4-Difluorophenoxy)methyl]-3,5-dimethyl-4-nitro-1H-pyrazole
Catalog # APOSPC410287-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.231 g/mol
Storage Temperature:Ambient
CAS Number:1001519-18-1
Formula:C₁₂H₁₁F₂N₃O₃
MDL Number:MFCD05861659