
2-(2,2,2-Trifluoroethoxy)ethanamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1035041-75-8
:
Formula:C₄H₈F₃NO·ClH
MDL Number:MFCD03196899
:179.569 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together