1-(2,2,2-Trifluoroethyl)piperazine dihydrochloride
Catalog # APOSPC410104-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:241.082 g/mol
Storage Temperature:Ambient
CAS Number:13349-91-2
Formula:C₆H₁₁F₃N₂·2ClH
MDL Number:MFCD08261052