2-Amino-4-(4-fluorophenyl)thiophene-3-carboxamide
Catalog # APOSPC410102-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:236.265 g/mol
Storage Temperature:Ambient
CAS Number:61019-24-7
Formula:C₁₁H₉FN₂OS
MDL Number:MFCD02855014