Order Entry
Italy
ContactUsLinkComponent
1-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine hydrochloride
1-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine hydrochloride
Catalog # APOSPC410086-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-amine hydrochloride
Catalog # APOSPC410086-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :243.657 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₂F₃N₃·ClH
MDL Number:MFCD25371115