2-(2,2,2-Trifluoroethoxy)aniline hydrochloride
Catalog # APOSPC410081-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.611 g/mol
Storage Temperature:Ambient
CAS Number:62149-83-1
Formula:C₈H₈F₃NO·ClH
MDL Number:MFCD11100362