Order Entry
Italy
ContactUsLinkComponent
4-(2,2-Difluoroethoxy)-2-methylaniline hydrochloride
4-(2,2-Difluoroethoxy)-2-methylaniline hydrochloride
Catalog # APOSPC410075-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(2,2-Difluoroethoxy)-2-methylaniline hydrochloride
Catalog # APOSPC410075-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.647 g/mol
Storage Temperature:Ambient
CAS Number:1431965-01-3
Formula:C₉H₁₁F₂NO·ClH
MDL Number:MFCD16621777