Order Entry
Italy
ContactUsLinkComponent
1-(2,5-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
1-(2,5-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Catalog # APOSPC410051-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,5-Difluorophenyl)-N-[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Catalog # APOSPC410051-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :251.275 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₁₅F₂N₃
MDL Number:MFCD08697228