Order Entry
Italy
ContactUsLinkComponent
2-(4-Fluorophenyl)-5-methylpyrazolo[1,5-a]pyrimidine-7-carbohydrazide
2-(4-Fluorophenyl)-5-methylpyrazolo[1,5-a]pyrimidine-7-carbohydrazide
Catalog # APOSPC410031-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Fluorophenyl)-5-methylpyrazolo[1,5-a]pyrimidine-7-carbohydrazide
Catalog # APOSPC410031-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :285.276 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₂FN₅O
MDL Number:MFCD06739368