Order Entry
Italy
ContactUsLinkComponent
N-t-BOC-5,5-Difluoropiperidine-3-Methanol
N-t-BOC-5,5-Difluoropiperidine-3-Methanol
Catalog # APOSPC409052-250MG
Supplier:  Apollo Scientific
CAS Number:  
N-t-BOC-5,5-Difluoropiperidine-3-Methanol
Catalog # APOSPC409052-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :251.27 g/mol
Storage Temperature:Ambient
CAS Number:1262412-64-5
Formula:C₁₁H₁₉F₂NO₃
MDL Number:MFCD16875553