2-Bromo-4-(pentafluoroethyl)pyridine
Catalog # APOSPC408839-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Tipo di conf.:Bottle
- Conf:1 g
- Seq:3047
Specifications
About this item
:
:276.001 g/mol
Storage Temperature:Ambiente
CAS Number:1816286-31-3
Formula:C₇H₃BrF₅N
MDL Number:MFCD28122888