

2,2-Bis(trifluoromethyl)tetrachloro-1,3-dioxolane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64499-81-6
:
Formula:C₅Cl₄F₆O₂
MDL Number:MFCD16619610
:347.855 g/mol
Boiling Pt:56 °C (25 mm Hg)
UN:1992
ADR:3+6.1,III
:GHS07
:GHS02
Storage Temperature:Ambient
Frequently Bought Together