Order Entry
Italy
ContactUsLinkComponent
(S)-4-(oxiran-2-yl)-2,8-bis(trifluoromethyl)quinoline
(S)-4-(oxiran-2-yl)-2,8-bis(trifluoromethyl)quinoline
Catalog # APOSPC404626-1G
Supplier:  Apollo Scientific
CAS Number:  
(S)-4-(oxiran-2-yl)-2,8-bis(trifluoromethyl)quinoline
Catalog # APOSPC404626-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :307.191 g/mol
Storage Temperature:Ambient
CAS Number:1292282-19-9
Formula:C₁₃H₇F₆NO
MDL Number:MFCD28580130