4-Bromo-3-chloro-5,8-difluoro-2-methylquinoline
Catalog # APOSPC404616-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:292.507 g/mol
Storage Temperature:Ambient
CAS Number:1333252-68-8
Formula:C₁₀H₅BrClF₂N
MDL Number:MFCD20036486