Order Entry
Italy
ContactUsLinkComponent
4-Bromo-3-chloro-5,7-difluoroquinoline
4-Bromo-3-chloro-5,7-difluoroquinoline
Catalog # APOSPC404605-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-3-chloro-5,7-difluoroquinoline
Catalog # APOSPC404605-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.481 g/mol
Storage Temperature:Ambient
CAS Number:1209122-08-6
Formula:C₉H₃BrClF₂N
MDL Number:MFCD13619734