3,4-Dichloro-5,7-difluoroquinoline
Catalog # APOSPC404600-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:234.03 g/mol
Storage Temperature:Ambient
CAS Number:1204810-53-6
Formula:C₉H₃Cl₂F₂N
MDL Number:MFCD13192995