Order Entry
Italy
ContactUsLinkComponent
2-Chloro-6-fluoroquinoline-3-carboxaldehyde
2-Chloro-6-fluoroquinoline-3-carboxaldehyde
Catalog # APOSPC404558-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-6-fluoroquinoline-3-carboxaldehyde
Catalog # APOSPC404558-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.604 g/mol
Storage Temperature:Ambient
CAS Number:749920-54-5
Formula:C₁₀H₅ClFNO
MDL Number:MFCD06337019